Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001115007.2 | 5406 | UTR 3 | NP_001108479.1 | ||
NM_001115008.2 | 5406 | UTR 3 | NP_001108480.1 | ||
NM_001288996.1 | 5406 | UTR 3 | NP_001275925.1 | ||
NM_001288997.1 | 5406 | UTR 3 | NP_001275926.1 | ||
NM_194282.3 | 5406 | UTR 3 | NP_919258.2 | ||
XM_005262750.4 | 5406 | Intron | XP_005262807.1 | ||
XM_006714081.3 | 5406 | Intron | XP_006714144.1 | ||
XM_017007728.1 | 5406 | Intron | XP_016863217.1 | ||
XM_017007729.1 | 5406 | Intron | XP_016863218.1 | ||
XM_017007730.1 | 5406 | Intron | XP_016863219.1 |