Product Details

SNP ID
rs7754671
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109296841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCATCCGATTAAAGAAGCTCCA[A/G]AGCACTCTTAGGTCCTTTCCTTTTA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CCDC162P PubMed Links

Gene Details

Gene
CCDC162P
Gene Name
coiled-coil domain containing 162, pseudogene
There are no transcripts associated with this gene.

View Full Product Details