Product Details

SNP ID
rs7308208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57060063 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTTCCACAAAGTCAGCAAATCTG[A/G]AAAAGAGAAGATAATACTCGTTAGA
Phenotype
MIM: 601478 MIM: 616496
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO1A PubMed Links

Gene Details

Gene
MYO1A
Gene Name
myosin IA
There are no transcripts associated with this gene.

Gene
NEMP1
Gene Name
nuclear envelope integral membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130963.1 Intron NP_001124435.1
NM_015257.2 Intron NP_056072.2
XM_011538056.2 Intron XP_011536358.1
XM_011538057.2 Intron XP_011536359.1
XM_011538058.2 Intron XP_011536360.1
XM_011538059.1 Intron XP_011536361.1
XM_011538060.2 Intron XP_011536362.1
XM_017019077.1 Intron XP_016874566.1
XM_017019078.1 Intron XP_016874567.1
XM_017019079.1 Intron XP_016874568.1

View Full Product Details