Product Details

SNP ID
rs6794014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:12798053 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGTAACTTAACCCCCATGAGTCT[A/C]CCTGTGCTTCTGTTTCTATAAAATG
Phenotype
MIM: 610403
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAND2 PubMed Links

Gene Details

Gene
CAND2
Gene Name
cullin associated and neddylation dissociated 2 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162499.1 Intron NP_001155971.1
NM_012298.2 Intron NP_036430.1
XM_011533503.2 Intron XP_011531805.1
XM_011533504.2 Intron XP_011531806.1

View Full Product Details