Product Details

SNP ID
rs3775998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:131643639 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATACACTTTTGGAAGGGAGGG[A/G]CTCTTAGAAAAAAAGAGAATAATTT
Phenotype
MIM: 610594 MIM: 610499
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FNIP1 PubMed Links

Gene Details

Gene
FNIP1
Gene Name
folliculin interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008738.2 4605 UTR 3 NP_001008738.2
NM_133372.2 4605 UTR 3 NP_588613.2
Gene
RAPGEF6
Gene Name
Rap guanine nucleotide exchange factor 6
There are no transcripts associated with this gene.

View Full Product Details