Product Details

SNP ID
rs6998258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66127154 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGCCTGAAACAATGTCCTCCACC[A/G]AGAGAAACGTAAAGGACACTTGATC
Phenotype
MIM: 606469
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM55 PubMed Links

Gene Details

Gene
TRIM55
Gene Name
tripartite motif containing 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033058.2 112 UTR 5 NP_149047.2
NM_184085.1 112 UTR 5 NP_908973.1
NM_184086.1 112 UTR 5 NP_908974.1
NM_184087.1 112 UTR 5 NP_908975.1
XM_005251316.4 112 UTR 5 XP_005251373.1
XM_011517613.2 112 UTR 5 XP_011515915.1
XM_011517614.2 112 UTR 5 XP_011515916.1
XM_011517615.2 112 UTR 5 XP_011515917.1
XM_011517616.2 112 UTR 5 XP_011515918.1
XM_011517617.2 112 Intron XP_011515919.1
XM_017013908.1 112 Intron XP_016869397.1
XM_017013909.1 112 Intron XP_016869398.1

View Full Product Details