Product Details

SNP ID
rs7788540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123816043 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCATAGCATTATGACAAAAATTTG[C/G]AAGAATCTCATGGGCACTAACCCAT
Phenotype
MIM: 604510
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HYAL4 PubMed Links

Gene Details

Gene
HYAL4
Gene Name
hyaluronoglucosaminidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012269.2 Intron NP_036401.2
XM_011515990.2 Intron XP_011514292.1
XM_011515991.2 Intron XP_011514293.1
XM_017011911.1 Intron XP_016867400.1
Gene
HYALP1
Gene Name
hyaluronoglucosaminidase pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details