Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001012454.5 | 445 | Intron | NP_001012457.3 | ||
NM_001128926.3 | 445 | Intron | NP_001122398.1 | ||
NM_001290254.1 | 445 | Intron | NP_001277183.1 | ||
NM_001290255.1 | 445 | Intron | NP_001277184.1 | ||
XM_011516128.2 | 445 | Intron | XP_011514430.1 | ||
XM_011516129.2 | 445 | Intron | XP_011514431.1 | ||
XM_011516130.2 | 445 | Intron | XP_011514432.1 | ||
XM_011516131.2 | 445 | Intron | XP_011514433.1 | ||
XM_011516132.2 | 445 | Intron | XP_011514434.1 | ||
XM_011516134.2 | 445 | Intron | XP_011514436.1 | ||
XM_011516135.2 | 445 | UTR 5 | XP_011514437.1 | ||
XM_011516136.2 | 445 | UTR 5 | XP_011514438.1 | ||
XM_017012147.1 | 445 | Intron | XP_016867636.1 | ||
XM_017012149.1 | 445 | Intron | XP_016867638.1 |