Product Details

SNP ID
rs6567573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:3622833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCATTATTTTGACATGCTCCCC[A/C]CTGCCAAAAAAAAATCAAGGTTATC
Phenotype
MIM: 300083
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRKX PubMed Links

Gene Details

Gene
PRKX
Gene Name
protein kinase, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005044.4 Intron NP_005035.1

View Full Product Details