Product Details

SNP ID
rs7128759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59807263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGTGATTATGTAGGCTGCAGGA[A/T]GAGGACTTTGGAATATCACTCTGGC
Phenotype
MIM: 611829 MIM: 600876
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPL16 PubMed Links

Gene Details

Gene
MRPL16
Gene Name
mitochondrial ribosomal protein L16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017840.3 Intron NP_060310.1
Gene
STX3
Gene Name
syntaxin 3
There are no transcripts associated with this gene.

View Full Product Details