Product Details

SNP ID
rs7973389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49586962 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGGCATCAGGACCCTCTCCCCA[C/T]TGGGGATCTGTAACTGACACAGGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM186B PubMed Links

Gene Details

Gene
FAM186B
Gene Name
family with sequence similarity 186 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032130.2 Intron NP_115506.1
XM_006719625.2 Intron XP_006719688.1
XM_006719626.2 Intron XP_006719689.1
XM_006719627.3 Intron XP_006719690.1
XM_011538796.2 Intron XP_011537098.1
XM_011538797.2 Intron XP_011537099.1
XM_017020008.1 Intron XP_016875497.1
Gene
LOC100422296
Gene Name
Parkinsonism associated deglycase pseudogene
There are no transcripts associated with this gene.

View Full Product Details