Product Details

SNP ID
rs7890232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:104106058 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATGTTCAGAGGCTCCCTGGCTTC[A/G]GCTCACTGCGTGCCAGTAGCACCAG
Phenotype
MIM: 300941
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A53 PubMed Links

Gene Details

Gene
SLC25A53
Gene Name
solute carrier family 25 member 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012755.4 Intron NP_001012773.2
XM_005262129.4 Intron XP_005262186.1
XM_011530952.2 Intron XP_011529254.1
XM_011530953.2 Intron XP_011529255.1
XM_017029514.1 Intron XP_016885003.1
XM_017029515.1 Intron XP_016885004.1
XM_017029516.1 Intron XP_016885005.1
Gene
ZCCHC18
Gene Name
zinc finger CCHC-type containing 18
There are no transcripts associated with this gene.

View Full Product Details