Product Details

SNP ID
rs7056971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55058835 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAAAACCGGAGAACAGTCTGGAG[C/T]GTTAGTCTTCGAAAATTTTATTGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAGE2B PubMed Links

Gene Details

Gene
PAGE2B
Gene Name
PAGE family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015038.2 Intron NP_001015038.1
XM_011530785.2 Intron XP_011529087.1
XM_011530786.2 Intron XP_011529088.1
XM_011530787.2 Intron XP_011529089.1
XM_017029513.1 Intron XP_016885002.1

View Full Product Details