Product Details

SNP ID
rs7934729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2404870 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGTTCCGCTGGAGGTCGGCAAAG[A/G]TCAGTGCACAACGTGCAGGGTGGCC
Phenotype
MIM: 186845 MIM: 604600 MIM: 603852
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD81 PubMed Links

Gene Details

Gene
CD81
Gene Name
CD81 molecule
There are no transcripts associated with this gene.

Gene
TRPM5
Gene Name
transient receptor potential cation channel subfamily M member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014555.3 3574 UTR 3 NP_055370.1
XM_017017628.1 3574 Intron XP_016873117.1
Gene
TSSC4
Gene Name
tumor suppressing subtransferable candidate 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297658.1 3574 Intron NP_001284587.1
NM_001297659.1 3574 Intron NP_001284588.1
NM_001297660.1 3574 Intron NP_001284589.1
NM_001297661.1 3574 Intron NP_001284590.1
NM_005706.3 3574 Intron NP_005697.2
XM_006718118.2 3574 Intron XP_006718181.1
XM_011519830.2 3574 Intron XP_011518132.1

View Full Product Details