Product Details
- SNP ID
-
rs6967578
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:150570406 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATTTCCCAGCTCCTGGTGGTCTC[A/C]CTATAACCAACAAATTAAACTGGCT
- Phenotype
-
MIM: 608087
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GIMAP4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111610351] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GIMAP4
- Gene Name
- GTPase, IMAP family member 4
- Gene
- LOC107986860
- Gene Name
- endogenous retrovirus group K member 19 Env polyprotein-like
There are no transcripts associated with this gene.
View Full Product Details