Product Details

SNP ID
rs7217182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37617544 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTAATATTCTCATAAAACTCAGC[C/T]AAGCTAGCACAAATTTACCTAAGTA
Phenotype
MIM: 612500 MIM: 607291
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX52 PubMed Links

Gene Details

Gene
DDX52
Gene Name
DEAD-box helicase 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291476.1 Intron NP_001278405.1
NM_007010.4 Intron NP_008941.3
XM_011524232.2 Intron XP_011522534.1
XM_011524233.2 Intron XP_011522535.1
Gene
MIR378J
Gene Name
microRNA 378j
There are no transcripts associated with this gene.

Gene
SYNRG
Gene Name
synergin, gamma
There are no transcripts associated with this gene.

View Full Product Details