Product Details

SNP ID
rs6708242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96341164 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGGATGATTCTTTGGAACATTCT[A/C]TTTGTTTTACATGTTTATGTTGTTT
Phenotype
MIM: 602332
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NCAPH PubMed Links

Gene Details

Gene
NCAPH
Gene Name
non-SMC condensin I complex subunit H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281710.1 Intron NP_001268639.1
NM_001281711.1 Intron NP_001268640.1
NM_001281712.1 Intron NP_001268641.1
NM_015341.4 Intron NP_056156.2
XM_005263908.3 Intron XP_005263965.1
XM_006712388.3 Intron XP_006712451.1

View Full Product Details