Product Details

SNP ID
rs8014867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70045009 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTAGCCCTTTGTCGTCACACACC[A/G]TTCCTATTTTTGTCCCATCTCTTCT
Phenotype
MIM: 607991
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC8A3 PubMed Links

Gene Details

Gene
SLC8A3
Gene Name
solute carrier family 8 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130417.2 2173 UTR 3 NP_001123889.1
NM_033262.4 2173 UTR 3 NP_150287.1
NM_058240.3 2173 UTR 3 NP_489479.1
NM_182932.2 2173 UTR 3 NP_891977.1
NM_182936.2 2173 UTR 3 NP_891981.1
NM_183002.2 2173 UTR 3 NP_892114.1
XM_006720240.3 2173 UTR 3 XP_006720303.1
XM_017021606.1 2173 UTR 3 XP_016877095.1
XM_017021607.1 2173 UTR 3 XP_016877096.1
XM_017021608.1 2173 UTR 3 XP_016877097.1
XM_017021609.1 2173 UTR 3 XP_016877098.1
XM_017021610.1 2173 Intron XP_016877099.1
XM_017021611.1 2173 UTR 3 XP_016877100.1

View Full Product Details