Product Details

SNP ID
rs7474580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:23205920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTTAAAAGACTAATGACATTTC[A/C]CAGGTAATGCAATTTAGAAAGAAAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C10orf67 PubMed Links

Gene Details

Gene
C10orf67
Gene Name
chromosome 10 open reading frame 67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153714.2 Intron NP_714925.2
XM_011519440.1 Intron XP_011517742.1
XM_011519441.1 Intron XP_011517743.1
XM_011519442.2 Intron XP_011517744.1
XM_011519443.1 Intron XP_011517745.1
XM_011519444.2 Intron XP_011517746.1
XM_011519445.2 Intron XP_011517747.1
XM_011519446.1 Intron XP_011517748.1
XM_017016029.1 Intron XP_016871518.1
XM_017016030.1 Intron XP_016871519.1
XM_017016031.1 Intron XP_016871520.1

View Full Product Details