Product Details
- SNP ID
-
rs6957219
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:101619046 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACACCAGGGGCGGGAGGTGCTCAC[A/G]GCCTCACCTCCTGGGCCCGGCCCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYL10
PubMed Links
Gene Details
- Gene
- MYL10
- Gene Name
- myosin light chain 10
View Full Product Details