Product Details

SNP ID
rs8092199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:20962255 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATCTTATAAAAGCAAAAAAGTT[A/T]TTGCCAGAAATTTTCCCATTAGTGT
Phenotype
MIM: 601702
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ROCK1 PubMed Links

Gene Details

Gene
ROCK1
Gene Name
Rho associated coiled-coil containing protein kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005406.2 Intron NP_005397.1
XM_011526137.2 Intron XP_011524439.1

View Full Product Details