Product Details

SNP ID
rs6882210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:122142745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGATGTCAAATGGTATGACAATC[A/G]TATTTTGATGACTTATTCACATAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100505841 PubMed Links

Gene Details

Gene
LOC100505841
Gene Name
zinc finger protein 474-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195535.1 Intron NP_001182464.1
XM_011543086.2 Intron XP_011541388.1
XM_017008912.1 Intron XP_016864401.1
Gene
ZNF474
Gene Name
zinc finger protein 474
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207317.2 Intron NP_997200.1

View Full Product Details