Product Details

SNP ID
rs8127192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34184691 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCACTTCACACATTCACAAATCAC[A/G]CACTTCTAGTTGGCTTTCTTGTGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00310 PubMed Links

Gene Details

Gene
LINC00310
Gene Name
long intergenic non-protein coding RNA 310
There are no transcripts associated with this gene.

View Full Product Details