Product Details
- SNP ID
-
rs7564675
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:135788554 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGTGGGTCCAGCATCTAGGAGAG[A/T]GTGACACAGGGTGGTTGGTGCTCTG
- Phenotype
-
MIM: 603202
MIM: 611216
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LCT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142752338] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LCT
- Gene Name
- lactase
- Gene
- UBXN4
- Gene Name
- UBX domain protein 4
There are no transcripts associated with this gene.
View Full Product Details