Product Details

SNP ID
rs7723416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:58587167 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTATAGCAAAAATTCACTTTTGT[A/G]GTATAAGAATGCTTTTCCATTGAAA
Phenotype
MIM: 612829
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB3C PubMed Links

Gene Details

Gene
RAB3C
Gene Name
RAB3C, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317915.1 Intron NP_001304844.1
NM_138453.3 Intron NP_612462.1

View Full Product Details