Product Details
- SNP ID
-
hCV29251163
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:113752870 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAGGGAGGGCAAATGTTATTTGG[A/G]AGTGTCCATGAGCATGAATGTGGTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLEKHS1
PubMed Links
Gene Details
- Gene
- PLEKHS1
- Gene Name
- pleckstrin homology domain containing S1
There are no transcripts associated with this gene.
View Full Product Details