Product Details

SNP ID
rs7637711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49792220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGTATGACAGACTCACAGTTTC[A/G]AGCCCTGAGCTTATCTACCCTCCTC
Phenotype
MIM: 606991
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDHR4 PubMed Links

Gene Details

Gene
CDHR4
Gene Name
cadherin related family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007540.3 Intron NP_001007541.2
XM_011533700.2 Intron XP_011532002.1
XM_011533701.2 Intron XP_011532003.1
XM_017006366.1 Intron XP_016861855.1
XM_017006367.1 Intron XP_016861856.1
XM_017006368.1 Intron XP_016861857.1
XM_017006369.1 Intron XP_016861858.1
XM_017006370.1 Intron XP_016861859.1
XM_017006371.1 Intron XP_016861860.1
Gene
IP6K1
Gene Name
inositol hexakisphosphate kinase 1
There are no transcripts associated with this gene.

View Full Product Details