Product Details

SNP ID
rs7481466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55979415 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATGGAATTCTTAGAATGGGGAAG[A/G]CAACTGTATAGTAACACAAAAGGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR7E5P PubMed Links

Gene Details

Gene
OR7E5P
Gene Name
olfactory receptor family 7 subfamily E member 5 pseudogene
There are no transcripts associated with this gene.

View Full Product Details