Product Details
- SNP ID
-
rs7645816
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:138951853 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCCTCCACTTCTCACTCAGTGCT[C/T]CCCACCAGGATGGGCTTAATGCCTT
- Phenotype
-
MIM: 605597
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FOXL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200854858] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FOXL2
- Gene Name
- forkhead box L2
There are no transcripts associated with this gene.
- Gene
- FOXL2NB
- Gene Name
- FOXL2 neighbor
- Gene
- LINC01391
- Gene Name
- long intergenic non-protein coding RNA 1391
There are no transcripts associated with this gene.
View Full Product Details