Product Details

SNP ID
rs7099154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:44297302 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCCCACCTTCCTTTGGATCCCTT[A/T]TAAGTAATCCAGGTCATAGATGATT
Phenotype
MIM: 600835
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C10orf142 PubMed Links

Gene Details

Gene
C10orf142
Gene Name
chromosome 10 open reading frame 142
There are no transcripts associated with this gene.

Gene
CXCL12
Gene Name
C-X-C motif chemokine ligand 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000609.6 Intron NP_000600.1
NM_001033886.2 Intron NP_001029058.1
NM_001178134.1 Intron NP_001171605.1
NM_001277990.1 Intron NP_001264919.1
NM_199168.3 Intron NP_954637.1

View Full Product Details