Product Details

SNP ID
rs8064613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75949381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCCTAAGATTAAAAAGAAAACA[C/T]CAGAGTTTGAGAACCTTGGAAAAAG
Phenotype
MIM: 609751
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOX1 PubMed Links

Gene Details

Gene
ACOX1
Gene Name
acyl-CoA oxidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185039.1 Intron NP_001171968.1
NM_004035.6 Intron NP_004026.2
NM_007292.5 Intron NP_009223.2
XM_011524868.2 Intron XP_011523170.1
XM_011524869.2 Intron XP_011523171.1

View Full Product Details