Product Details

SNP ID
rs6434369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:189771100 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAAATTAGAACACTTGACTCAGA[A/G]GCTGCTTGTAAGACACTTGGCTATC
Phenotype
MIM: 609803 MIM: 610073
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKAR PubMed Links

Gene Details

Gene
ANKAR
Gene Name
ankyrin and armadillo repeat containing
There are no transcripts associated with this gene.

Gene
ORMDL1
Gene Name
ORMDL sphingolipid biosynthesis regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128150.1 1349 UTR 3 NP_001121622.1
NM_016467.4 1349 UTR 3 NP_057551.1
XM_005246970.2 1349 UTR 3 XP_005247027.1
XM_011512199.2 1349 Intron XP_011510501.1
XM_011512200.2 1349 Intron XP_011510502.1
XM_017005347.1 1349 UTR 3 XP_016860836.1
XM_017005348.1 1349 Intron XP_016860837.1
Gene
OSGEPL1
Gene Name
O-sialoglycoprotein endopeptidase-like 1
There are no transcripts associated with this gene.

Gene
OSGEPL1-AS1
Gene Name
OSGEPL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details