Product Details

SNP ID
rs6430353
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:148884275 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCAGACAGAGACAGAGGCCAAGA[C/T]TGTAGTCTTAGAGAAAAATAGGATG
Phenotype
MIM: 604593
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KIF5C PubMed Links

Gene Details

Gene
KIF5C
Gene Name
kinesin family member 5C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004522.2 Intron NP_004513.1
XM_011511157.2 Intron XP_011509459.1
XM_017004062.1 Intron XP_016859551.1
Gene
LOC101928553
Gene Name
uncharacterized LOC101928553
There are no transcripts associated with this gene.

View Full Product Details