Product Details
- SNP ID
-
rs7023002
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:107485637 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGATGACTCAGTTGGGAACTTGA[A/C]CATGATTGTAGTGCTTTCTGGCTGG
- Phenotype
-
MIM: 602253
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KLF4
PubMed Links
Gene Details
- Gene
- KLF4
- Gene Name
- Kruppel like factor 4
View Full Product Details