Product Details

SNP ID
rs7122879
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:121026586 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAGTTAAAACTTCATCAGTGGGG[C/T]CACATTCTTGATCAAATGCTTTACG
Phenotype
MIM: 610451
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TBCEL PubMed Links

Gene Details

Gene
TBCEL
Gene Name
tubulin folding cofactor E like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130047.1 Intron NP_001123519.1
NM_152715.3 Intron NP_689928.3
XM_017017326.1 Intron XP_016872815.1

View Full Product Details