Product Details

SNP ID
rs8112382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7731069 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGACCTCCTCCTTCAGGGCACCCAG[A/C]GCCGCCGTCTGCTTCGAGGCTGGAA
Phenotype
MIM: 604672 MIM: 616256
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CD209 PubMed Links

Gene Details

Gene
CD209
Gene Name
CD209 molecule
There are no transcripts associated with this gene.

Gene
CLEC4G
Gene Name
C-type lectin domain family 4 member G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244856.1 309 Silent Mutation GCG,GCT A68A NP_001231785.1
NM_198492.3 309 Silent Mutation GCG,GCT A80A NP_940894.1

View Full Product Details