Product Details

SNP ID
rs7109250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72190626 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTCCCTGAGAGTACAGATTTCTC[C/T]GGCGTGGCCCTCAAGGTTAGTGAGT
Phenotype
MIM: 136430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOLR1 PubMed Links

Gene Details

Gene
FOLR1
Gene Name
folate receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000802.3 253 Intron NP_000793.1
NM_016724.2 253 UTR 5 NP_057936.1
NM_016725.2 253 Intron NP_057937.1
NM_016729.2 253 Intron NP_057941.1

View Full Product Details