Product Details
- SNP ID
-
hCV29370670
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:72192617 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTGGAGTGACCCATCCTGGGGTTG[A/C]TTGATTTAGGGATTTAGACTGGGAA
- Phenotype
-
MIM: 136430
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FOLR1
PubMed Links
Gene Details
- Gene
- FOLR1
- Gene Name
- folate receptor 1
There are no transcripts associated with this gene.
View Full Product Details