Product Details

SNP ID
rs6141897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33236859 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTCTTTGGTCTCCTGGTTCTTC[G/T]CATGCAATGAAGACTTTGATGGCCC
Phenotype
MIM: 607412
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BPIFA1 PubMed Links

Gene Details

Gene
BPIFA1
Gene Name
BPI fold containing family A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243193.1 Intron NP_001230122.1
NM_016583.3 Intron NP_057667.1
NM_130852.2 Intron NP_570913.1
Gene
BPIFA3
Gene Name
BPI fold containing family A member 3
There are no transcripts associated with this gene.

View Full Product Details