Product Details

SNP ID
rs6119651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31671443 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTGGTGCCTTTGCCAACACTGTC[A/C]TTTAACTGGTCACTCCATATCTTCC
Phenotype
MIM: 600039
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BCL2L1 PubMed Links

Gene Details

Gene
BCL2L1
Gene Name
BCL2 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191.3 Intron NP_001182.1
NM_001317919.1 Intron NP_001304848.1
NM_001317920.1 Intron NP_001304849.1
NM_001317921.1 Intron NP_001304850.1
NM_001322239.1 Intron NP_001309168.1
NM_001322240.1 Intron NP_001309169.1
NM_001322242.1 Intron NP_001309171.1
NM_138578.2 Intron NP_612815.1
XM_011528964.2 Intron XP_011527266.1
XM_017027993.1 Intron XP_016883482.1

View Full Product Details