Product Details

SNP ID
rs7177328
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52013267 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATCCTATCAAGAAAGGAAGTGGT[A/G]TTTCTTTGTAATGACTCACTTGAAG
Phenotype
MIM: 602904
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MAPK6 PubMed Links

Gene Details

Gene
MAPK6
Gene Name
mitogen-activated protein kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002748.3 Intron NP_002739.1
XM_005254537.2 Intron XP_005254594.1
XM_005254538.2 Intron XP_005254595.1
XM_005254539.3 Intron XP_005254596.1
XM_011521782.1 Intron XP_011520084.1

View Full Product Details