Product Details

SNP ID
rs8069233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:67072632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGAGCTCCTTCACCCTACTGC[A/G]GAAGAGCAGTGCAGACCCCAGCAGG
Phenotype
MIM: 606699
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HELZ PubMed Links

Gene Details

Gene
HELZ
Gene Name
helicase with zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014877.3 11636 UTR 3 NP_055692.2
XM_005257888.4 11636 UTR 3 XP_005257945.1
XM_005257889.4 11636 UTR 3 XP_005257946.1
XM_006722214.3 11636 UTR 3 XP_006722277.1
XM_006722215.3 11636 UTR 3 XP_006722278.1
XM_006722216.3 11636 UTR 3 XP_006722279.1
XM_011525544.2 11636 UTR 3 XP_011523846.1
XM_017025477.1 11636 UTR 3 XP_016880966.1
XM_017025478.1 11636 UTR 3 XP_016880967.1

View Full Product Details