Product Details

SNP ID
rs6969516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27746019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAAAGTATAAGGTTGGATTGTGG[A/G]TTTTTAAAAAACTGTTATAATTGAG
Phenotype
MIM: 605326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAX1BP1 PubMed Links

Gene Details

Gene
TAX1BP1
Gene Name
Tax1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079864.2 Intron NP_001073333.1
NM_001206901.1 Intron NP_001193830.1
NM_001206902.1 Intron NP_001193831.1
NM_006024.6 Intron NP_006015.4
XM_005249900.2 Intron XP_005249957.1
XM_017012754.1 Intron XP_016868243.1

View Full Product Details