Product Details

SNP ID
rs6910022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:47235405 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTGGTGTCTTTAATAAAAAGAAG[A/C]GATGAGGACACAGACGCACACAGAG
Phenotype
MIM: 605732
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TNFRSF21 PubMed Links

Gene Details

Gene
TNFRSF21
Gene Name
TNF receptor superfamily member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014452.4 Intron NP_055267.1
XM_017010744.1 Intron XP_016866233.1

View Full Product Details