Product Details

SNP ID
rs7743255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:50818191 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAAACTGAGGTGTATTTAAGTTT[C/T]CAAAGGTTTTAGGTAAAATGACCAG
Phenotype
MIM: 601601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TFAP2B PubMed Links

Gene Details

Gene
TFAP2B
Gene Name
transcription factor AP-2 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003221.3 500 Intron NP_003212.2
XM_011514837.2 500 Intron XP_011513139.1
XM_017011233.1 500 UTR 5 XP_016866722.1
XM_017011234.1 500 Intron XP_016866723.1
XM_017011235.1 500 Intron XP_016866724.1

View Full Product Details