Product Details

SNP ID
rs8077233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16549143 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTTATGATTTTTAAAACTTTCT[A/G]TATATTTGAAAACTCATAATAAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF287 PubMed Links

Gene Details

Gene
ZNF287
Gene Name
zinc finger protein 287
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020653.2 5659 Intron NP_065704.2
XM_011523968.2 5659 UTR 3 XP_011522270.1
XM_017024887.1 5659 UTR 3 XP_016880376.1

View Full Product Details