Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001322405.1 | 4592 | UTR 3 | NP_001309334.1 | ||
NM_001322406.1 | 4592 | UTR 3 | NP_001309335.1 | ||
NM_001322408.1 | 4592 | UTR 3 | NP_001309337.1 | ||
NM_001322410.1 | 4592 | UTR 3 | NP_001309339.1 | ||
NM_001322412.1 | 4592 | UTR 3 | NP_001309341.1 | ||
NM_001322413.1 | 4592 | UTR 3 | NP_001309342.1 | ||
NM_001322414.1 | 4592 | UTR 3 | NP_001309343.1 | ||
NM_001322415.1 | 4592 | UTR 3 | NP_001309344.1 | ||
NM_001322416.1 | 4592 | UTR 3 | NP_001309345.1 | ||
NM_001322417.1 | 4592 | Intron | NP_001309346.1 | ||
NM_001322418.1 | 4592 | Intron | NP_001309347.1 | ||
NM_001322419.1 | 4592 | Intron | NP_001309348.1 | ||
NM_015491.2 | 4592 | UTR 3 | NP_056306.1 | ||
NM_032870.3 | 4592 | UTR 3 | NP_116259.2 | ||
XM_005266912.3 | 4592 | Intron | XP_005266969.2 | ||
XM_017010710.1 | 4592 | Intron | XP_016866199.1 | ||
XM_017010711.1 | 4592 | Intron | XP_016866200.1 | ||
XM_017010712.1 | 4592 | Intron | XP_016866201.1 | ||
XM_017010713.1 | 4592 | Intron | XP_016866202.1 |