Product Details

SNP ID
rs10209670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169853154 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGTGAGAGGGAGTAGTTACTTA[C/T]GAATGCTTGAGAAAATGGTGCTGTC
Phenotype
MIM: 613831
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBR3 PubMed Links

Gene Details

Gene
UBR3
Gene Name
ubiquitin protein ligase E3 component n-recognin 3 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172070.3 Intron NP_742067.3
XM_005246299.4 Intron XP_005246356.1
XM_005246300.4 Intron XP_005246357.1
XM_006712268.3 Intron XP_006712331.1
XM_006712269.3 Intron XP_006712332.1
XM_006712270.3 Intron XP_006712333.2
XM_006712271.3 Intron XP_006712334.1
XM_011510604.2 Intron XP_011508906.1
XM_017003356.1 Intron XP_016858845.1
XM_017003357.1 Intron XP_016858846.1
XM_017003358.1 Intron XP_016858847.1
XM_017003359.1 Intron XP_016858848.1

View Full Product Details