Product Details

SNP ID
rs10414591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:22417320 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAGGGAACAACAGACACTGAGGC[A/G]TAGTTGAGGTTAGAGGACAAAGAGG
Phenotype
MIM: 603980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF98 PubMed Links

Gene Details

Gene
ZNF98
Gene Name
zinc finger protein 98
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098626.1 Intron NP_001092096.1
XM_011527705.1 Intron XP_011526007.1
XM_017026373.1 Intron XP_016881862.1

View Full Product Details