Product Details

SNP ID
rs9603823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113308822 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTTTTTTGGCCCACGTTAGTATA[C/T]TGCTTCACTGTAGTTCATTTGACCT
Phenotype
MIM: 153330
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAMP1 PubMed Links

Gene Details

Gene
LAMP1
Gene Name
lysosomal associated membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005561.3 Intron NP_005552.3
XM_011537494.1 Intron XP_011535796.1

View Full Product Details